Merck
  • Plant-like biosynthetic pathways in bacteria: from benzoic acid to chalcone.
Plant-like biosynthetic pathways in bacteria: from benzoic acid to chalcone.
Journal of natural products (2002-12-28)
Bradley S Moore, Christian Hertweck, Jörn N Hopke, Miho Izumikawa, John A Kalaitzis, George Nilsen, Thomas O'Hare, Jörn Piel, Paul R Shipley, Longkuan Xiang, Michael B Austin, Joseph P Noel
ABSTRACT
Although phenylpropanoids and flavonoids are common plant natural products, these major classes of biologically active secondary metabolites are largely absent from bacteria. The ubiquitous plant enzymes phenylalanine ammonia-lyase (PAL) and chalcone synthase (CHS) are key biosynthetic catalysts in phenylpropanoid and flavonoid assembly, respectively. Until recently, few bacterial counterparts were known, thus reflecting the dearth of these plant natural products in bacteria. This review highlights our progress on the biochemical and genetic characterization of recently identified streptomycete biosynthetic pathways to benzoic acid and type III polyketide synthase (PKS)-derived products. The sediment-derived bacterium "Streptomyces maritimus" produces benzoyl-CoA in a plant-like manner from phenylalanine involving a PAL-mediated reaction through cinnamic acid during the biosynthesis of the polyketide antibiotic enterocin. All but one of the genes encoding benzoyl-CoA biosynthesis in "S. maritimus" have been cloned, sequenced, and inactivated, providing a model for benzoate biosynthesis not only in this bacterium, but in plants where benzoic acid is an important constituent of many products. The recent discovery that bacteria harbor homodimeric PKSs belonging to the plant CHS superfamily of condensing enzymes has further linked the biosynthetic capabilities of plants and bacteria. A bioinformatics approach led to the prediction that the model actinomycete Streptomyces coelicolor A3(2) contains up to three type III PKSs. Biochemical analysis of one of the recombinant type III PKSs from S. coelicolor demonstrated activity as a 1,3,6,8-tetrahydroxynaphthalene synthase (THNS). A homology model of THNS based upon the known three-dimensional structure of CHS was constructed to explore the structural and mechanistic details of this new subclass of bacterial PKSs.
Product Description
Supelco
Mettler-Toledo Calibration substance ME 18555, Benzoic acid, analytical standard, (for the calibration of the melting point system), traceable to primary standards (LGC)
Benzoic acid, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Benzoic acid, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99.9% (alkalimetric)
Sigma-Aldrich
Benzoic acid, ≥99.5%, FCC, FG
Sigma-Aldrich
Benzoic acid, natural, ≥99.5%, FCC, FG
Supelco
Benzoic acid, reference material for titrimetry, certified by BAM, >99.5%
Sigma-Aldrich
Benzoic acid, ACS reagent, ≥99.5%
Sigma-Aldrich
Benzoic acid, ReagentPlus®, 99%
Sigma-Aldrich
Benzoic acid, purified by sublimation, ≥99%
Sigma-Aldrich
Benzoic acid, meets analytical specification of Ph. Eur., BP, USP, FCC, E210, 99.5-100.5% (alkalimetric)
Supelco
Benzoic acid, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Melting point standard 121-123°C, analytical standard
Sigma-Aldrich
Benzoic acid, SAJ special grade, ≥99.5%
Sigma-Aldrich
Benzoic acid, SAJ first grade, ≥99.5%
Supelco
Benzoic acid, Standard for quantitative NMR, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
USP
Benzoic acid, United States Pharmacopeia (USP) Reference Standard