Merck
Methimazole embryopathy: delineation of the phenotype.
American journal of medical genetics (1999-03-17)
M Clementi, E Di Gianantonio, E Pelo, I Mammi, R T Basile, R Tenconi
ABSTRACT
We report on a further case of congenital anomalies in a child exposed to methimazole during the first trimester of pregnancy (from first to seventh gestational week), and define a specific malformation pattern related to prenatal methimazole exposure and consisting of choanal and esophageal atresia, scalp defects, minor facial anomalies and psychomotor delay.
Product Description
Supelco
Methimazole, VETRANAL®, analytical standard
Sigma-Aldrich
2-Mercapto-1-methylimidazole, ≥99%
Supelco
Methimazole, analytical standard
USP
Methimazole, United States Pharmacopeia (USP) Reference Standard
Thiamazole, European Pharmacopoeia (EP) Reference Standard