Merck
  • Development of novel chiral urea catalysts for the hetero-Michael reaction.
Development of novel chiral urea catalysts for the hetero-Michael reaction.
Chemical & pharmaceutical bulletin (2004-04-02)
Yoshihiro Sohtome, Aya Tanatani, Yuichi Hashimoto, Kazuo Nagasawa
ABSTRACT
Chiral urea compounds 10a-g were synthesized as catalysts for conjugate addition of pyrrolidine (2) to gamma-crotonolactone (3). In the presence of a catalytic amount of the chiral ureas, this hetero-Michael reaction was greatly accelerated. Asymmetric induction was observed with the catalysts 10e, 10f, and 10g.
Sigma-Aldrich
2(5H)-Furanone, 98%
Pricing and availability is not currently available.