Merck
  • Asymmetric synthesis of α-allyl-α-aryl α-amino acids by tandem alkylation/π-allylation of α-iminoesters.
Asymmetric synthesis of α-allyl-α-aryl α-amino acids by tandem alkylation/π-allylation of α-iminoesters.
Organic letters (2014-03-29)
John M Curto, Joshua S Dickstein, Simon Berritt, Marisa C Kozlowski
ABSTRACT
The first asymmetric synthesis of α-allyl-α-aryl α-amino acids by means of a three-component coupling of α-iminoesters, Grignard reagents, and cinnamyl acetate is reported. Notably, the enolate from the tandem process provides a much higher level of reactivity and selectivity than the same enolate generated via direct deprotonation, presumably due to differences in the solvation/aggregation state. A novel method for removal of a homoallylic amine protecting group delivers the free amine congeners. The α-allyl group offers a means to generate further valuable α-amino acid structures as exemplified by ring closing metathesis to generate a higher ring homologue of α-aryl-proline.
Sigma-Aldrich
L-Proline, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
L-Proline, 99%, FCC, FG
SAFC
L-Proline
Sigma-Aldrich
L-Proline, from non-animal source, meets EP, USP testing specifications, suitable for cell culture
Sigma-Aldrich
L-Proline, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
L-Proline, SAJ special grade, ≥99.0%
Supelco
L-Proline, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
L-Proline, Pharmaceutical Secondary Standard; Certified Reference Material
USP
L-Proline, United States Pharmacopeia (USP) Reference Standard
Proline, European Pharmacopoeia (EP) Reference Standard