Merck
  • A large-scale cluster randomized trial to determine the effects of community-based dietary sodium reduction--the China Rural Health Initiative Sodium Reduction Study.
A large-scale cluster randomized trial to determine the effects of community-based dietary sodium reduction--the China Rural Health Initiative Sodium Reduction Study.
American heart journal (2013-11-02)
Nicole Li, Lijing L Yan, Wenyi Niu, Darwin Labarthe, Xiangxian Feng, Jingpu Shi, Jianxin Zhang, Ruijuan Zhang, Yuhong Zhang, Hongling Chu, Andrea Neiman, Michael Engelgau, Paul Elliott, Yangfeng Wu, Bruce Neal
ABSTRACT
Cardiovascular diseases are the leading cause of death and disability in China. High blood pressure caused by excess intake of dietary sodium is widespread and an effective sodium reduction program has potential to improve cardiovascular health. This study is a large-scale, cluster-randomized, trial done in five Northern Chinese provinces. Two counties have been selected from each province and 12 townships in each county making a total of 120 clusters. Within each township one village has been selected for participation with 1:1 randomization stratified by county. The sodium reduction intervention comprises community health education and a food supply strategy based upon providing access to salt substitute. Subsidization of the price of salt substitute was done in 30 intervention villages selected at random. Control villages continued usual practices. The primary outcome for the study is dietary sodium intake level estimated from assays of 24-hour urine. The trial recruited and randomized 120 townships in April 2011. The sodium reduction program was commenced in the 60 intervention villages between May and June of that year with outcome surveys scheduled for October to December 2012. Baseline data collection shows that randomisation achieved good balance across groups. The establishment of the China Rural Health Initiative has enabled the launch of this large-scale trial designed to identify a novel, scalable strategy for reduction of dietary sodium and control of blood pressure. If proved effective, the intervention could plausibly be implemented at low cost in large parts of China and other countries worldwide.
Sigma-Aldrich
Sodium, SAJ first grade, ≥99.5%
Sigma-Aldrich
Sodium, CP
Sigma-Aldrich
Sodium, ACS reagent, dry
Sigma-Aldrich
Sodium, cubes, contains mineral oil, 99.9% trace metals basis
Sigma-Aldrich
Sodium, 99.95% trace metals basis, ingot
Sigma-Aldrich
Sodium, 25-35 wt % dispersion in paraffin
Sigma-Aldrich
Sodium, in kerosene, pieces (large), ≥99.8% (sodium basis)