Merck
  • A new protocol for ecotoxicological assessment of seawater using nauplii of Tisbe biminiensis (Copepoda:Harpacticoida).
A new protocol for ecotoxicological assessment of seawater using nauplii of Tisbe biminiensis (Copepoda:Harpacticoida).
Ecotoxicology and environmental safety (2013-06-19)
Beatriz R B O Lavorante, Deloar D Oliveira, Bruno V M Costa, Lília P Souza-Santos
ABSTRACT
Copepods are largely used in toxicity tests. The nauplii of these organisms are more sensitive to contaminants than the adult stage. The aim of the present study was to test a protocol for the use of nauplii of the copepod Tisbe biminiensis in the ecotoxicological assessment of seawater. The sensitivity of these organisms to zinc sulphate (ZnSO4·7H2O) was also determined. The following conditions were established for the protocol based on the best development of nauplii to copepodites: 72-h duration, the microalga Chaetocerus gracilis at 2.5×10(5)cellsmL(-1) as feed and incubation temperature of 28°C. In the zinc sulphate sensitivity tests, EC50-72 h and LC50/72 h were 3.25±0.59 mg L(-1) and 3.46±0.72 mg L(-1), respectively, as estimated by the final number of copepodites and total number of live animals in relation to the mean number of inoculated nauplii. The estimated NOEC was 2.0 mg L(-1). The test developed is fast and not labour intensive. T. biminiensis nauplii exhibit sensitivity to zinc sulphate similar to that of other species of copepods employed in water toxicity tests, demonstrating the usefulness of these organisms in ecotoxicological studies involving samples of environmental seawater.
Sigma-Aldrich
Zinc sulfate heptahydrate, SAJ first grade, ≥99.0%
Sigma-Aldrich
Zinc sulfate heptahydrate, BioUltra, for molecular biology, 2.0 M in H2O
Sigma-Aldrich
Zinc sulfate heptahydrate, ≥99.95% trace metals basis
Sigma-Aldrich
Zinc sulfate heptahydrate, meets USP testing specifications
Sigma-Aldrich
Zinc sulfate heptahydrate, suitable for plant cell culture
Sigma-Aldrich
Zinc sulfate heptahydrate, BioReagent, suitable for cell culture
Sigma-Aldrich
Zinc sulfate heptahydrate, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.5%
Sigma-Aldrich
Zinc sulfate heptahydrate, ACS reagent, 99%
Sigma-Aldrich
Zinc sulfate heptahydrate, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Zinc sulfate solution, 0.3 N