Merck
Silver-mediated fluorination of aryl silanes.
Tetrahedron (2011-06-22)
Pingping Tang, Tobias Ritter
ABSTRACT
A regiospecific silver-mediated fluorination of aryl silanes is reported. The reaction is operationally simple, and employs Ag(2)O as readily available, inexpensive silver source, which can be recovered.
Sigma-Aldrich
Bis(acetonitrile)(1,5-cyclooctadiene)rhodium(I)tetrafluoroborate
Pricing and availability is not currently available.
Sigma-Aldrich
1-Fluoro-3-nitrobenzene, 97%
Pricing and availability is not currently available.