Merck
  • γ-Radiation promotes immunological recognition of cancer cells through increased expression of cancer-testis antigens in vitro and in vivo.
γ-Radiation promotes immunological recognition of cancer cells through increased expression of cancer-testis antigens in vitro and in vivo.
PloS one (2011-12-06)
Anu Sharma, Beata Bode, Roland H Wenger, Kuno Lehmann, Alessandro A Sartori, Holger Moch, Alexander Knuth, Lotta von Boehmer, Maries van den Broek
ABSTRACT
γ-radiation is an effective treatment for cancer. There is evidence that radiotherapy supports tumor-specific immunity. It was described that irradiation induces de novo protein synthesis and enhances antigen presentation, we therefore investigated whether γ-radiation results in increased expression of cancer-testis (CT) antigens and MHC-I, thus allowing efficient immunological control. This is relevant because the expression of CT-antigens and MHC-I on tumor cells is often heterogeneous. We found that the changes induced by γ-radiation promote the immunological recognition of the tumor, which is illustrated by the increased infiltration by lymphocytes after radiotherapy. We compared the expression of CT-antigens and MHC-I in various cancer cell lines and fresh biopsies before and after in vitro irradiation (20 Gy). Furthermore, we compared paired biopsies that were taken before and after radiotherapy from sarcoma patients. To investigate whether the changed expression of CT-antigens and MHC-I is specific for γ-radiation or is part of a generalized stress response, we analyzed the effect of hypoxia, hyperthermia and genotoxic stress on the expression of CT-antigens and MHC-I. In vitro irradiation of cancer cell lines and of fresh tumor biopsies induced a higher or de novo expression of different CT-antigens and a higher expression of MHC-I in a time- and dose-dependent fashion. Importantly, we show that irradiation of cancer cells enhances their recognition by tumor-specific CD8+ T cells. The analysis of paired biopsies taken from a cohort of sarcoma patients before and after radiotherapy confirmed our findings and, in addition showed that irradiation resulted in higher infiltration by lymphocytes. Other forms of stress did not have an impact on the expression of CT-antigens or MHC-I. Our findings suggest that γ-radiation promotes the immunological recognition of the tumor. We therefore propose that combining radiotherapy with treatments that support tumor specific immunity may result in increased therapeutic efficacy.
Sigma-Aldrich
Propidium iodide, ≥94% (HPLC)
Sigma-Aldrich
Propidium iodide, ≥94.0% (HPLC)
Sigma-Aldrich
Fetal Bovine Serum, Australia origin, γ-irradiated, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Fetal Bovine Serum, USA origin, γ-irradiated by SER-TAIN process, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Fetal Bovine Serum, Australia origin, USDA approved, Heat Inactivated, suitable for cell culture
Sigma-Aldrich
Fetal Bovine Serum, non-USA origin, from USDA approved countries, Heat Inactivated, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Fetal Bovine Serum, USA origin, Heat Inactivated, sterile-filtered, suitable for cell culture
SAFC
Fetal Bovine Serum, Australia origin, γ-irradiated, suitable for cell culture, suitable for hybridoma
Sigma-Aldrich
Fetal Bovine Serum, USA origin, Dialyzed by ultrafiltration against 0.15 M NaCl, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Fetal Bovine Serum, USA origin, sterile-filtered, suitable for cell culture, suitable for hybridoma
Sigma-Aldrich
Fetal Bovine Serum, USDA approved, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Fetal Bovine Serum, suitable for, Australia origin, USDA approved, sterile-filtered, suitable for cell culture, suitable for hybridoma
Sigma-Aldrich
Fetal Bovine Serum, USA origin, Heat Inactivated, sterile-filtered, suitable for cell culture, suitable for insect cell culture, suitable for hybridoma
Sigma-Aldrich
Fetal Bovine Serum, USA origin, Charcoal Stripped, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Fetal Bovine Serum, Canada origin, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Fetal Bovine Serum, Australia origin, USDA approved, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Fetal Bovine Serum, USA origin, suitable for cell culture
Sigma-Aldrich
Etoposide, synthetic, 95.0-105.0%, powder