Merck
  • Can serum beta-hydroxybutyrate be used to diagnose diabetic ketoacidosis?
Can serum beta-hydroxybutyrate be used to diagnose diabetic ketoacidosis?
Diabetes care (2008-01-11)
Mae Sheikh-Ali, Brad S Karon, Ananda Basu, Yogish C Kudva, Lisa A Muller, Jia Xu, W Frederick Schwenk, John M Miles
ABSTRACT
Current criteria for the diagnosis of diabetic ketoacidosis (DKA) are limited by their nonspecificity (serum bicarbonate [HCO(3)] and pH) and qualitative nature (the presence of ketonemia/ketonuria). The present study was undertaken to determine whether quantitative measurement of a ketone body anion could be used to diagnose DKA. A retrospective review of records from hospitalized diabetic patients was undertaken to determine the concentration of serum beta-hydroxybutyrate (betaOHB) that corresponds to a HCO(3) level of 18 mEq/l, the threshold value for diagnosis in recently published consensus criteria. Simultaneous admission betaOHB and HCO(3) values were recorded from 466 encounters, 129 in children and 337 in adults. A HCO(3) level of 18 mEq/l corresponded with betaOHB levels of 3.0 and 3.8 mmol/l in children and adults, respectively. With the use of these threshold betaOHB values to define DKA, there was substantial discordance (approximately > or = 20%) between betaOHB and conventional diagnostic criteria using HCO(3), pH, and glucose. In patients with DKA, there was no correlation between HCO(3) and glucose levels on admission and a significant but weak correlation between betaOHB and glucose levels (P < 0.001). Where available, serum betaOHB levels > or = 3.0 and > or = 3.8 mmol/l in children and adults, respectively, in the presence of uncontrolled diabetes can be used to diagnose DKA and may be superior to the serum HCO(3) level for that purpose. The marked variability in the relationship between betaOHB and HCO(3) is probably due to the presence of other acid-base disturbances, especially hyperchloremic, nonanion gap acidosis.
Sigma-Aldrich
Acetone, suitable for residue analysis, ≥99.5%
Sigma-Aldrich
1-Butanol, SAJ first grade, ≥99.0%
Sigma-Aldrich
Acetone, suitable for residue analysis, JIS 5000
Sigma-Aldrich
Acetone, suitable for HPLC
Sigma-Aldrich
1-Butanol, JIS special grade, ≥99.0%
Sigma-Aldrich
Acetone, ≥99.5%, suitable for residue analysis
Sigma-Aldrich
3-Hydroxybutyric acid, 95%
Supelco
Glycerin, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
1-Butanol, suitable for HPLC, 99.8%
Sigma-Aldrich
Acetone, ≥99%, FCC, FG
Sigma-Aldrich
Dimethylamine, anhydrous, ≥99%
Sigma-Aldrich
Dodecanedioic acid, 99%
Sigma-Aldrich
Acetone, natural, ≥97%
Sigma-Aldrich
1-Butanol, suitable for HPLC
Sigma-Aldrich
Acetone, suitable for chromatography, ≥99.8%
Sigma-Aldrich
Acetone, JIS special grade, ≥99.5%
Sigma-Aldrich
Acetone, SAJ first grade, ≥99.0%
Sigma-Aldrich
1-Butanol, Molecular Biology, ≥99%
Sigma-Aldrich
Chromium, powder, ≥99% trace metals basis, <45 μm
Supelco
1-Butanol, analytical standard
Supelco
Acetone, analytical standard
Sigma-Aldrich
Glycerol solution, 83.5-89.5% (T)
Sigma-Aldrich
Pyruvaldehyde solution, 40 wt. % in H2O
Sigma-Aldrich
Acetone, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Chromium, chips, 99.995% trace metals basis
Sigma-Aldrich
Chromium, chips, thickness ~2 mm, 99.5%
Sigma-Aldrich
1-Butanol, anhydrous, 99.8%
Sigma-Aldrich
Chromium, powder, 99.5%, −100 mesh
Sigma-Aldrich
Acetone, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.5% (GC)
Sigma-Aldrich
1-Butanol, ACS reagent, ≥99.4%