Merck
  • Improvement on the freeze-thaw stability of corn starch gel by the polysaccharide from leaves of Corchorus olitorius L.
Improvement on the freeze-thaw stability of corn starch gel by the polysaccharide from leaves of Corchorus olitorius L.
Carbohydrate polymers (2013-04-03)
Eiji Yamazaki, Toru Sago, Yoshiaki Kasubuchi, Kazuhito Imamura, Toshio Matsuoka, Osamu Kurita, Hironobu Nambu, Yasuki Matsumura
ABSTRACT
Effect of the polysaccharide from leaves of Corchorus olitorius L. (PLC) on the freeze-thaw (FT) stability of corn starch gel was studied. PLC was incorporated into the starch gel at 0.7% and total solid was adjusted to 6.0%. The syneresis was measured by the centrifugal-filtration method and, as a result, addition of PLC reduced effectively the syneresis of the starch gel even after 5 FT cycles, which was less than one third that of the normal starch gel. The rheological changes of the starch/PLC gel during the FT treatments were evaluated while the gel remained on the rheometer plate. The starch/PLC gel had less significant changes in the rheological parameters during the FT cycles than starch/guar gum or xanthan gum gel systems. SEM images showed that PLC stabilized the gel matrix surrounding pores, which would contribute to both a lower syneresis production and a higher stability in the rheological behavior at FT.
Sigma-Aldrich
Starch from potato, suitable for electrophoresis
Sigma-Aldrich
Starch from potato, Soluble
Sigma-Aldrich
Starch from potato, Powder
Sigma-Aldrich
Starch from wheat, Unmodified
Sigma-Aldrich
Starch, from potato, tested according to Ph. Eur.
Sigma-Aldrich
Starch from corn, practical grade
Supelco
Starch from corn, for use with Total Dietary Fiber Control Kit, TDF-C10
Supelco
Starch from corn, analytical standard, analytical standard for Starch Assay Kits SA-20 and STA-20
Sigma-Aldrich
Starch from corn
Sigma-Aldrich
Starch from rice
Sigma-Aldrich
Starch, puriss. p.a., from potato, reag. ISO, reag. Ph. Eur., soluble
Sigma-Aldrich
Starch from corn