Merck
  • Dermal pharmacokinetics of Terpinen-4-ol following topical administration of Zingiber cassumunar (plai) oil.
Dermal pharmacokinetics of Terpinen-4-ol following topical administration of Zingiber cassumunar (plai) oil.
Planta medica (2012-10-03)
Kotchaphan Chooluck, Rajendra P Singh, Korbtham Sathirakul, Hartmut Derendorf
ABSTRACT
The purpose of this study was to investigate dermal pharmacokinetics of terpinen-4-ol in rats following topical administration of plai oil derived from the rhizomes of Zingiber cassumunar Roxb. Unbound terpinen-4-ol concentrations in dermal tissue were measured by microdialysis. The dermal pharmacokinetic study of terpinen-4-ol was performed under non-occlusive conditions. The oil was topically applied at a dose of 2, 4, and 8 mg/cm2 plai oil corresponding to the amount of 1.0, 1.9, and 3.8 mg/cm2 terpinen-4-ol, respectively. Following topical application of the oil, terpinen-4-ol rapidly distributed into the dermis and demonstrated linear pharmacokinetics with no changes in the dose-normalized area under the concentration-time curves across the investigated dosage range. The mean percentages of free terpinen-4-ol distributed in the dermis per amount of administered were 0.39 ± 0.06 %, 0.41 ± 0.08 %, and 0.30 ± 0.03 % for 2, 4, and 8 mg/cm2 doses, respectively. The dermal pharmacokinetics of terpinen-4-ol could provide information for its further formulation development and therapy schedules.
Product Description
Terpinen 4-ol, primary reference standard
Sigma-Aldrich
4-Carvomenthenol, ≥95%, FCC, FG
Sigma-Aldrich
4-Carvomenthenol, natural, ≥95%, FG