Merck
  • Development and validation of a liquid chromatographic/electrospray ionization mass spectrometric method for the quantitation of prazepam and its main metabolites in human plasma.
Development and validation of a liquid chromatographic/electrospray ionization mass spectrometric method for the quantitation of prazepam and its main metabolites in human plasma.
Journal of mass spectrometry : JMS (2005-02-16)
Paraskevi Valavani, Julia Atta-Politou, Irene Panderi
ABSTRACT
A method was developed and fully validated for the quantitation of prazepam and its major metabolites, oxazepam and nordiazepam, in human plasma. Sample pretreatment was achieved by solid-phase extraction using Oasis HLB cartridges. The extracts were analysed by high-performance liquid chromatography (HPLC) coupled with single-quadrupole mass spectrometry (MS) with an electrospray ionization interface. The MS system was operated in the selected ion monitoring mode. HPLC was performed isocratically on a reversed-phase XTerra MS C18 analytical column (150 x 3.0 mm i.d., particle size 5 microm). Diazepam was used as the internal standard for quantitation. The assay was linear over a concentration range of 5.0-1000 ng ml(-1) for all compounds analyzed. The limit of quantitation was 5 ng ml(-1) for all compounds. Quality control samples (5, 10, 300 and 1000 ng ml(-1)) in five replicates from three different runs of analysis demonstrated an intra-assay precision (CV) of < or = 9.1%, an inter-assay precision of < or = 6.0% and an overall accuracy (relative error) of < 4.6%. The method can be used to quantify prazepam and its metabolites in human plasma covering a variety of pharmacokinetic or bioequivalence studies.
Sigma-Aldrich
Prazepam
Pricing and availability is not currently available.
Supelco
Prazepam solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Pricing and availability is not currently available.