- Vibrational analysis of substituted benzonitriles. IV. Transferability of force constants--the case of some aminobenzonitriles.
Vibrational analysis of substituted benzonitriles. IV. Transferability of force constants--the case of some aminobenzonitriles.
A zero-order normal coordinate analysis was made for the n-plane vibrations of p-, m- and o-aminobenzonitrile and 2-chloro, 5-aminobenzonitrile by transferring the force constants from our earlier work. The observed and calculated frequencies agree with an average error of 19.3 cm-1. Unambiguous vibrational assignments of in-plane fundamentals of the four molecules have been made, and several assignments suggested by earlier workers have been revised.