Merck
  • Dearomatized white spirit inhalation exposure causes long-lasting neurophysiological changes in rats.
Dearomatized white spirit inhalation exposure causes long-lasting neurophysiological changes in rats.
Neurotoxicology and teratology (1996-01-01)
S P Lund, L Simonsen, U Hass, O Ladefoged, H R Lam, G Ostergaard
ABSTRACT
Exposure for 6 h per day, 5 days per week, during a period of 6 months to the organic solvent dearomatized white spirit (0, 400, and 800 ppm) was studied in rats that were 3 months old when the repeated exposure was initiated. After an exposure-free period of 2-6 months duration, neurophysiological, neurobehavioral, and macroscopic pathologic examinations were performed. The study revealed exposure-related changes in sensory evoked potentials and a decrease in motor activity during dark (no light) periods but no white spirit-induced changes in learning and memory functions. The measurements of the flash evoked potential (FEP), somatosensory evoked potential (SEP), and auditory brain stem response (ABR) all demonstrated dose-dependent increases of the amplitudes of the early latency peaks of the sensory evoked potentials (EPs). Furthermore, an increase of the dose showed that the measurements of FEP and SEP revealed changes in the later-latency peaks, which reflect the more associative aspects of sensory processing. The results demonstrated that 6 months of exposure to dearomatized white spirit induced long-lasting and possible irreversible effects in the nervous system of the rat.
Sigma-Aldrich
Petroleum ether, SAJ first grade, ≥90.0%
Sigma-Aldrich
Petroleum ether, anhydrous
Sigma-Aldrich
Petrol, SAJ first grade, ≥90.0%
Sigma-Aldrich
Petroleum ether, JIS special grade, ≥90.0%
Sigma-Aldrich
Petroleum ether, ACS reagent
Sigma-Aldrich
Petroleum ether, spectrophotometric grade
Sigma-Aldrich
Petroleum ether, ACS reagent
Sigma-Aldrich
Petroleum ether, ACS reagent
Sigma-Aldrich
Ligroin, SAJ first grade, ≥90.0%
Sigma-Aldrich
Ligroin, JIS special grade, ≥90.0%