Merck
  • Cross-linked hydrogel and polyester resorbable ventilation tubes in a Chinchilla model.
Cross-linked hydrogel and polyester resorbable ventilation tubes in a Chinchilla model.
The Laryngoscope (2013-03-21)
Albert H Park, David Hoyt, David Britt, Shane Chase, Kristina Tansavatdi, Lisa Hunter, Lawrence McGill, Xiaoming Sheng, Aleksander Skardal, Glenn D Prestwich
ABSTRACT
To determine the resorption rate and biocompatibility characteristics of novel cross-linked hydrogel ventilation tubes and varied formulations of polyester ventilation tubes in a Chinchilla model. Animal Study. Three cross-linked glycosaminoglycan hydrogel ventilation tubes fabricated by cross-linking thiol-modified chondroitin sulfate or thiol-modified carboxymethylated hyaluronic acid, four different polyester ventilation tubes (poly L-lactide [PLA], 50/50 poly D,L-lactide-co-glycolide [PLGA], and silver-impregnated versions of PLA and PLGA tubes) were placed into the tympanic membranes of chinchillas. Commercially available fluoroplastic ventilation tubes were placed in the contralateral ear of each animal to serve as a control. Integrity of the tubes was assessed by weekly otoscopy. Biocompatibility was assessed by auditory brainstem response, by otoscopic and histologic examination of the tympanic membrane at the tube site. The hydrogel tubes had very short resorption times that expanded and enlarged the myringotomy site. PLGA and silver-coated PLGA tubes maintained their integrity in the tympanic membrane for similar durations of 18.9 ± 6.4 days and 21.0 ± 6.0 days, respectively. The silver-coated PLGA tubes had lower neutrophil and fibrosis scores than PLGA tubes. PLA tubes demonstrated equivalent findings to commercially available nonresorbable tubes with respect to otoscopic findings, auditory brainstem response thresholds, and histologic inflammatory scores. Resorbable polyester pressure equalization tubes demonstrate predictable resorption behavior and similar biocompatibility characteristics when compared with nonresorbable tubes. Silver modification may confer some stability to PLGA tubes. Hydrogel tubes have very short resorption times, tend to enlarge the myringotomy site, and show greater inflammatory changes.
Sigma-Aldrich
Poly(ethylene glycol) dimethacrylate, average Mn 2000, contains ~1000 ppm MeHQ as stabilizer
Sigma-Aldrich
Poly(ethylene glycol) dimethacrylate, average Mn 6,000, contains 1000 ppm 4-methoxyphenol as inhibitor
Sigma-Aldrich
Poly(ethylene glycol) dimethacrylate, average Mn 20,000, contains MEHQ as inhibitor
Sigma-Aldrich
Poly(ethylene glycol) dimethacrylate, average Mn 750, contains 900-1100 ppm MEHQ as inhibitor
Sigma-Aldrich
Poly(ethylene glycol) dimethacrylate, average Mn 550, contains 80-120 ppm MEHQ as inhibitor, 270-330 ppm BHT as inhibitor
Sigma-Aldrich
Poly(ethylene glycol) dimethacrylate, average Mn 10,000, contains MEHQ as inhibitor