Merck
Surface-enhanced Raman spectroscopy applied to food safety.
Annual review of food science and technology (2013-01-10)
Ana Paula Craig, Adriana S Franca, Joseph Irudayaraj
ABSTRACT
Surface-enhanced Raman spectroscopy (SERS) is an advanced Raman technique that enhances the vibrational spectrum of molecules adsorbed on or in the vicinity of metal particles and/or surfaces. Because of its readiness, sensitivity, and minimum sample preparation requirements, SERS is being considered as a powerful technique for food inspection. Key aspects of food-safety assurance, spectroscopy methods, and SERS are briefly discussed in an extended introduction of this review. The recent and potential advances in SERS are highlighted in sections that deal with the (a) detection of food-borne pathogenic microorganisms and (b) the detection of food contaminants and adulteration, concentrated specifically on antibiotics, drugs, hormones, melamine, and pesticides. This review provides an outlook of the work done and a perspective on the future directions of SERS as a reliable tool for food-safety assessment.
Product Description
Supelco
Melamine, Pharmaceutical Secondary Standard; Certified Reference Material
Pricing and availability is not currently available.
Sigma-Aldrich
Melamine, 99%
Pricing and availability is not currently available.
Supelco
Melamine, analytical standard
Pricing and availability is not currently available.