Merck
  • Bromine radical-mediated sequential radical rearrangement and addition reaction of alkylidenecyclopropanes.
Bromine radical-mediated sequential radical rearrangement and addition reaction of alkylidenecyclopropanes.
Journal of the American Chemical Society (2012-12-28)
Takashi Kippo, Kanako Hamaoka, Ilhyong Ryu
ABSTRACT
Bromine radical-mediated cyclopropylcarbinyl-homoallyl rearrangement of alkylidenecyclopropanes was effectively accomplished by C-C bond formation with allylic bromides, which led to the syntheses of 2-bromo-1,6-dienes. A three-component coupling reaction comprising alkylidenecyclopropanes, allylic bromides, and carbon monoxide also proceeded well to give 2-bromo-1,7-dien-5-ones in good yield.
Sigma-Aldrich
Bromine water, CP
Sigma-Aldrich
Bromine, JIS special grade, ≥99.0%
Sigma-Aldrich
Propane, 99.5%
Sigma-Aldrich
Bromine, ≥99.99% trace metals basis
Sigma-Aldrich
Propane, 98%
Sigma-Aldrich
Bromine, SAJ first grade, ≥97.0%
Sigma-Aldrich
Bromine, reagent grade
Sigma-Aldrich
Bromine, ACS reagent, ≥99.5%