Merck
  • Discovery and preliminary structure-activity relationship analysis of 1,14-sperminediphenylacetamides as potent and selective antimalarial lead compounds.
Discovery and preliminary structure-activity relationship analysis of 1,14-sperminediphenylacetamides as potent and selective antimalarial lead compounds.
Bioorganic & medicinal chemistry letters (2012-12-26)
Lydia P P Liew, Marcel Kaiser, Brent R Copp
ABSTRACT
Screening of synthesized and isolated marine natural products for in vitro activity against four parasitic protozoa has identified the ascidian metabolite 1,14-sperminedihomovanillamide (orthidine F, 1) as being a non-toxic, moderate growth inhibitor of Plasmodium falciparum (IC(50) 0.89 μM). Preliminary structure-activity relationship investigation identified essentiality of the spermine polyamine core and the requirement for 1,14-disubstitution for potent activity. One analogue, 1,14-spermine-di-(2-hydroxyphenylacetamide) (3), exhibited two orders of magnitude increased anti-P. f activity (IC(50) 8.6 nM) with no detectable in vitro toxicity. The ease of synthesis of phenylacetamido-polyamines, coupled with potent nM levels of activity towards dual drug resistant strains of P. falciparum makes this compound class of interest in the development of new antimalarial therapeutics.
Sigma-Aldrich
Spermine, ≥99.0% (GC)
Sigma-Aldrich
Spermine, ≥97%
Sigma-Aldrich
Spermine, suitable for cell culture, BioReagent