Merck
  • Ultrastructural study of vitellogenesis and oogenesis of Metadena depressa (Stossich, 1883) Linton, 1910 (Digenea, Cryptogonimidae), intestinal parasite of Dentex dentex (Pisces, Teleostei).
Ultrastructural study of vitellogenesis and oogenesis of Metadena depressa (Stossich, 1883) Linton, 1910 (Digenea, Cryptogonimidae), intestinal parasite of Dentex dentex (Pisces, Teleostei).
Comptes rendus biologies (2012-12-04)
Samuel Greani, Yann Quilichini, Joséphine Foata, Zdzisław Swiderski, Bernard Marchand
ABSTRACT
The ultrastructural organization of the female reproductive system of Metadena depressa, digenean intestinal parasite of Sparidae (Dentex dentex), was investigated by electron microscopy. The vitellogenesis is divided into four stages: stage I, vitellocytes have a cytoplasm mainly filled with ribosomes and few mitochondria; stage II, beginning of the synthetic activity; stage III, active shell globule clusters synthesis; stage IV, mature vitellocytes are filled with shell globule clusters and generally contain several large lipid droplets. Glycogen granules are grouped at the periphery of the cell. The three stages of the oogenesis process take place in the ovary: stage I, oogonia are undifferentiated small cells located at the periphery of the organ; stage II, primary oocytes possess a higher nucleo-cytoplasmic ratio and a nucleus with a nucleolus and synaptonemal complexes indicating the zygotene-pachytene stage of the first meiotic division; stage III, mature oocytes are located in the proximal region of the organ and possess a cytoplasmic chromatoid body and cortical granules in a monolayer close to the periphery of the cell.
Sigma-Aldrich
Lead(II) acetate trihydrate, ≥99.99% trace metals basis
Sigma-Aldrich
Lead(II) acetate trihydrate, 99.999% trace metals basis
Sigma-Aldrich
Lead(II) acetate trihydrate, JIS special grade, ≥99.5%
Sigma-Aldrich
Lead(II) acetate trihydrate, ACS reagent, ≥99%
Sigma-Aldrich
Lead(II) acetate trihydrate, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., 99.5-102.0%