Merck
  • Copper- and phosphine-ligand-free palladium-catalyzed direct allylation of electron-deficient polyfluoroarenes with allylic chlorides.
Copper- and phosphine-ligand-free palladium-catalyzed direct allylation of electron-deficient polyfluoroarenes with allylic chlorides.
Chemistry (Weinheim an der Bergstrasse, Germany) (2012-10-12)
Yan-Bo Yu, Shilu Fan, Xingang Zhang
ABSTRACT
Simply id(all)ylic: a copper- and phosphine-ligand-free Pd-catalyzed direct allylation of electron-deficient polyfluoroarenes with allylic chlorides and the reaction mechanism are described. The simple catalytic system, broad substrate scope, and excellent functional-group compatibility of this protocol provides a useful and facile access to allylated polyfluoroarenes.
Sigma-Aldrich
Allyl chloride, ReagentPlus®, 99%
Sigma-Aldrich
Allyl chloride, reagent grade, 98%
SAFC
Allyl chloride