Merck
  • Production of p-xylene from biomass by catalytic fast pyrolysis using ZSM-5 catalysts with reduced pore openings.
Production of p-xylene from biomass by catalytic fast pyrolysis using ZSM-5 catalysts with reduced pore openings.
Angewandte Chemie (International ed. in English) (2012-10-06)
Yu-Ting Cheng, Zhuopeng Wang, Christopher J Gilbert, Wei Fan, George W Huber
ABSTRACT
Pores for thought: Chemical liquid deposition of silica onto ZSM-5 catalysts led to smaller pore openings that resulted in >90% selectivity for p-xylene over the other xylenes in the catalytic fast pyrolysis of furan and 2-methylfuran (see scheme). The p-xylene selectivity increased from 51% with gallium spray-dried ZSM-5 to 72% with a pore-mouth-modified catalyst in the pyrolysis of pine wood.
Sigma-Aldrich
p-Xylene, anhydrous, ≥99%
Sigma-Aldrich
p-Xylene, SAJ first grade, ≥99.0%
Supelco
p-Xylene, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
p-Xylene, analytical standard
Sigma-Aldrich
p-Xylene, ReagentPlus®, 99%
Sigma-Aldrich
p-Xylene, puriss. p.a., ≥99.0% (GC)
Sigma-Aldrich
p-Xylene, suitable for HPLC, ≥99%