Merck
  • Ginger extract and zingerone ameliorated trinitrobenzene sulphonic acid-induced colitis in mice via modulation of nuclear factor-κB activity and interleukin-1β signalling pathway.
Ginger extract and zingerone ameliorated trinitrobenzene sulphonic acid-induced colitis in mice via modulation of nuclear factor-κB activity and interleukin-1β signalling pathway.
Food chemistry (2012-09-29)
Chien-Yun Hsiang, Hsin-Yi Lo, Hui-Chi Huang, Chia-Cheng Li, Shih-Lu Wu, Tin-Yun Ho
ABSTRACT
Ginger is a commonly used spice with anti-inflammatory potential. Colitis is the common pathological lesion of inflammatory bowel diseases. In this study, we investigated the therapeutic effects of ginger and its component zingerone in mice with 2,4,6-trinitrobenzene sulphonic acid (TNBS)-induced colitis. Ginger and zingerone ameliorated TNBS-induced colonic injury in a dose-dependent manner. Pathway analysis of ginger- and zingerone-regulated gene expression profiles showed that ginger and zingerone significantly regulated cytokine-related pathways. Network analysis showed that nuclear factor-κB (NF-κB) and interleukin-1β (IL-1β) were key molecules involved in the expression of ginger- and zingerone-affected genes. Ex vivo imaging and immunohistochemical staining further verified that ginger and zingerone suppressed TNBS-induced NF-κB activation and IL-1β protein level in the colon. In conclusion, ginger improved TNBS-induced colitis via modulation of NF-κB activity and IL-1β signalling pathway. Moreover, zingerone might be the active component of ginger responsible for the amelioration of colitis induced by TNBS.
Sigma-Aldrich
Vanillylacetone, ≥96%, FG
Sigma-Aldrich
Guaiacol, natural, ≥99%, FG
Sigma-Aldrich
Guaiacol, oxidation indicator
Supelco
Guaiacol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Guaiacol, SAJ first grade, ≥98.0%