Merck
Acyl homoserine-lactone quorum-sensing signal generation.
Proceedings of the National Academy of Sciences of the United States of America (1999-04-14)
M R Parsek, D L Val, B L Hanzelka, J E Cronan, E P Greenberg
ABSTRACT
Acyl homoserine lactones (acyl-HSLs) are important intercellular signaling molecules used by many bacteria to monitor their population density in quorum-sensing control of gene expression. These signals are synthesized by members of the LuxI family of proteins. To understand the mechanism of acyl-HSL synthesis we have purified the Pseudomonas aeruginosa RhlI protein and analyzed the kinetics of acyl-HSL synthesis by this enzyme. Purified RhlI catalyzes the synthesis of acyl-HSLs from acyl-acyl carrier proteins and S-adenosylmethionine. An analysis of the patterns of product inhibition indicated that RhlI catalyzes signal synthesis by a sequential, ordered reaction mechanism in which S-adenosylmethionine binds to RhlI as the initial step in the enzymatic mechanism. Because pathogenic bacteria such as P. aeruginosa use acyl-HSL signals to regulate virulence genes, an understanding of the mechanism of signal synthesis and identification of inhibitors of signal synthesis has implications for development of quorum sensing-targeted antivirulence molecules.
Sigma-Aldrich
N-Heptanoyl-L-homoserine lactone, ≥96% (HPLC)
Sigma-Aldrich
N-Dodecanoyl-L-homoserine lactone, ≥96% (HPLC)
Sigma-Aldrich
N-(3-Hydroxydecanoyl)-DL-homoserine lactone
Sigma-Aldrich
N-(3-Hydroxyoctanoyl)-DL-homoserine lactone, ≥95% (sum of isomers, HPLC)
Sigma-Aldrich
N-(3-Hydroxytetradecanoyl)-DL-homoserine lactone, ≥96% (HPLC), carbon 64.7-67.3 %
Sigma-Aldrich
N-Decanoyl-L-homoserine lactone, ≥96% (HPLC)
Sigma-Aldrich
N-Hexanoyl-L-homoserine lactone, ≥96% (HPLC)
Sigma-Aldrich
N-Octanoyl-L-homoserine lactone, ≥96% (HPLC)
Sigma-Aldrich
N-(3-Hydroxydodecanoyl)-DL-homoserine lactone, ≥97% (HPLC)