Merck
  • The membrane binding kinetics of full-length PKCα is determined by membrane lipid composition.
The membrane binding kinetics of full-length PKCα is determined by membrane lipid composition.
Biochimica et biophysica acta (2012-07-31)
Angel Pérez-Lara, Antonio L Egea-Jiménez, Alessio Ausili, Senena Corbalán-García, Juan C Gómez-Fernández
ABSTRACT
Protein kinase Cα (PKCα) is activated by its translocation to the membrane. Activity assays show the importance of PIP(2) in determining the specific activity of this enzyme. A FRET stopped flow fluorescence study was carried out to monitor the rapid kinetics of protein binding to model membranes containing POPC/POPS/DOG and eventually PIP(2). The results best fitted a binding mechanism in which protein bound to the membrane following a two-phase mechanism with a first bimolecular reaction followed by a slow unimolecular reaction. In the absence of PIP(2), the rapid protein binding rate was especially dependent on POPS concentration. Formation of the slow high affinity complex during the second phase seems to involve specific interactions with POPS and DOG since it is only sensitive to changes within relatively low concentration ranges of these lipids. Both the association and dissociation rate constants fell in the presence of PIP(2). We propose a model in which PKCα binds to the membranes via a two-step mechanism consisting of the rapid membrane initial recruitment of PKCα driven by interactions with POPS and/or PIP(2) although interactions with DOG are involved too. PKCα searches on the lipid bilayer in two dimensions to establish interactions with its specific ligands.
Sigma-Aldrich
Dioleoylglycerol, ≥99%, mixture of 1,3- and 1,2-isomers, liquid
Pricing and availability is not currently available.