Merck
  • Removing endotoxin from plasmid samples by Triton X-114 isothermal extraction.
Removing endotoxin from plasmid samples by Triton X-114 isothermal extraction.
Analytical biochemistry (2012-03-01)
Rui Ma, Jun Zhao, Hui-Cong Du, Shuang Tian, Li-Wen Li
ABSTRACT
Triton X-114 temperature transition extraction has been considered to be a simple and cost-effective strategy to eliminate endotoxin from plasmid preparations. However, a repeated cooling-heating process may promote the degradation of plasmid DNA. Based on the finding that the cloud point of Triton X-114 solution increases substantially in the presence of small amounts of sodium dodecyl sulfate (SDS) and that electrolytes decrease the cloud point of Triton X-114-SDS solution drastically, we designed a Triton X-114 isothermal extraction method for removing endotoxin from plasmid samples and found that it has the same endotoxin removal efficiency when compared with the temperature transition extraction method.
Sigma-Aldrich
IGEPAL® CA-720, average Mn ~735
Sigma-Aldrich
IGEPAL® CA-630, for molecular biology
Sigma-Aldrich
IGEPAL® CA-630, viscous liquid
Sigma-Aldrich
Triton X-114, laboratory grade
Sigma-Aldrich
Triton X-165 solution, Non-ionic, CMC-0.43 mM, suitable for electrophoresis, cloud point >100 °C
Supelco
IGEPAL® CA-630
Sigma-Aldrich
Triton X-114
Sigma-Aldrich
Triton X-405 solution, 70% in H2O