Merck
  • Quantification of prothrombin in human plasma amplified by autocatalytic reaction.
Quantification of prothrombin in human plasma amplified by autocatalytic reaction.
Analytical chemistry (2012-02-14)
Ana Virel, Laura Saa, Valeri Pavlov
ABSTRACT
By site directed mutagenesis, we have produced recombinant mutants of human and mouse prethrombin-2 which are able to convert themselves autocatalytically into α-thrombin. We also have created a new method to amplify the signal of bioanalytical assays based on the autocatalytic activation of these mutated proenzymes. The activation of the mutants by active α-thrombin triggers an autocatalytic reaction which leads to more active thrombin resulting in the amplification of the readout signal. Addition of mutated mouse prethrombin-2 into the conventional assay for prothrombin level in human plasma, employing ecarin and the fluorogenic substrate, resulted in improvement of the detection limit by 2 orders of magnitude.
Sigma-Aldrich
Rhodamine 110 chloride, suitable for fluorescence, BioReagent, ≥99.0% (UV)
Sigma-Aldrich
Rhodamine 110 chloride, Dye content ≥75 %