Merck
Clinical inquiry. Which drugs work best for early Parkinson's disease?
The Journal of family practice (2012-02-09)
Liza Jain, Rebecca Benko, Sarah Safranek
ABSTRACT
Levodopa/carbidopa is the most effective medical therapy for Parkinson's disease, but it's associated with dyskinesia.
Sigma-Aldrich
Ropinirole hydrochloride, powder, ≥98% (HPLC)