Merck
  • Photochemistry of acetylacetone isolated in parahydrogen matrices upon 266 nm irradiation.
Photochemistry of acetylacetone isolated in parahydrogen matrices upon 266 nm irradiation.
Physical chemistry chemical physics : PCCP (2012-02-07)
Rolando R Lozada-García, Justinas Ceponkus, Michèle Chevalier, Wutharath Chin, Jean-Michel Mestdagh, Claudine Crépin
ABSTRACT
The photochemistry of the chelated enol form of acetylacetone (AcAc) was investigated by UV excitation of the S(2) state at 266 nm in parahydrogen matrices, complemented by experiments in neon and normal hydrogen matrices. Infrared (IR) spectroscopy, combined with theoretical calculations, was used to identify the photoproducts. Isomerization towards various non-chelated forms (no intramolecular H-bond) of AcAc is the dominant channel whereas fragmentation is very minor. The isomerization kinetics is monitored by IR spectroscopy. Among the seven non-chelated conformers of AcAc, only three are formed in parahydrogen matrices, whereas four are observed in normal hydrogen matrices. This difference suggests that an active tunnelling process between conformers occurs in parahydrogen but is quenched in normal hydrogen where guest-host interactions are stronger. Fragmentation and isomerization of excited AcAc are discussed in the light of these new data. The role of the intermediate triplet state in the S(2)→ S(0) relaxation is confirmed, as the importance of phonons in the condensed phase.
Product Description
Supelco
Acetylacetone, analytical standard
Sigma-Aldrich
Acetylacetone, JIS special grade, ≥99%
Sigma-Aldrich
Acetylacetone, ReagentPlus®, ≥99%
Sigma-Aldrich
Acetylacetone, produced by Wacker Chemie AG, Burghausen, Germany, ≥99.5% (GC)