Merck
  • New solid lipid microparticles for controlled ibuprofen release: formulation and characterization study.
New solid lipid microparticles for controlled ibuprofen release: formulation and characterization study.
International journal of pharmaceutics (2011-10-27)
Laurent Perge, Mike Robitzer, Coralie Guillemot, Jean-Marie Devoisselle, Françoise Quignard, Philippe Legrand
ABSTRACT
A hot melt dispersion method was used to prepare new sustained release ibuprofen composite microparticles of a solid lipid at ambient temperature, cetyl alcohol. The dispersion of colloidal silicon dioxide nanoparticles (hydrophilic Aerosil 200 or hydrophobic Aerosil R974) either in the oily phase or in the aqueous phase led to the preparation of large (about 400 μm diameter) surfactant free free-flowing particles. Mapping-scanning electronic microscopy using silicon probe revealed that silicon was in the oily core in all cases. The nature of silica nanoparticles and the way used for their dispersion influenced the internal structure of the composite microparticles and the aggregation of nanoparticles in the core of the microparticles. Hydrophobic Aerosil R974 allowed the formation of homogeneous microparticles. Although silica nanoparticles had no influence on thermic profile, crystalline state of ibuprofen and lipid, they had an influence on the kinetics drug release related to the increase of the size of the composite solid lipid microparticles prepared.
Sigma-Aldrich
1-Hexadecanol, 95%
Sigma-Aldrich
1-Hexadecanol, ReagentPlus®, 99%
Sigma-Aldrich
1-Hexadecanol, ≥99%
Supelco
Cetyl alcohol, analytical standard
Sigma-Aldrich
Cetyl alcohol, SAJ special grade, ≥98.0%
Cetyl alcohol, European Pharmacopoeia (EP) Reference Standard
Supelco
Cetyl Alcohol, Pharmaceutical Secondary Standard; Certified Reference Material