Merck
  • Biodegradation of sulfamethazine by Trametes versicolor: Removal from sewage sludge and identification of intermediate products by UPLC-QqTOF-MS.
Biodegradation of sulfamethazine by Trametes versicolor: Removal from sewage sludge and identification of intermediate products by UPLC-QqTOF-MS.
The Science of the total environment (2011-09-29)
Ma Jesús García-Galán, Carlos E Rodríguez-Rodríguez, Teresa Vicent, Gloria Caminal, M Silvia Díaz-Cruz, Damià Barceló
ABSTRACT
Degradation of the sulfonamide sulfamethazine (SMZ) by the white-rot fungus Trametes versicolor was assessed. Elimination was achieved to nearly undetectable levels after 20 h in liquid medium when SMZ was added at 9 mg L(-1). Experiments with purified laccase and laccase-mediators resulted in almost complete removal. On the other hand, inhibition of SMZ degradation was observed when piperonilbutoxide, a cytochrome P450-inhibitor, was added to the fungal cultures. UPLC-QqTOF-MS analysis allowed the identification and confirmation of 4 different SMZ degradation intermediates produced by fungal cultures or purified laccase: desulfo-SMZ, N4-formyl-SMZ, N4-hydroxy-SMZ and desamino-SMZ; nonetheless SMZ mineralization was not demonstrated with the isotopically labeled sulfamethazine-phenyl-13C6 after 7 days. Inoculation of T. versicolor to sterilized sewage sludge in solid-phase systems showed complete elimination of SMZ and also of other sulfonamides (sulfapyridine, sulfathiazole) at real environmental concentrations, making this fungus an interesting candidate for further remediation research.
Product Description
Supelco
Sulfamethazine, VETRANAL®, analytical standard
Sigma-Aldrich
Sulfamethazine, 99.0-101.0% (on dried basis)