Merck
  • Catalytic, asymmetric hypervinylogous Mukaiyama aldol reactions of extended furan-based silyl enolates.
Catalytic, asymmetric hypervinylogous Mukaiyama aldol reactions of extended furan-based silyl enolates.
Organic letters (2011-08-13)
Claudio Curti, Lucia Battistini, Andrea Sartori, Alessio Lodola, Marco Mor, Gloria Rassu, Giorgio Pelosi, Franca Zanardi, Giovanni Casiraghi
ABSTRACT
Virtually perfect transmittal of the enolate reactivity up to five conjugated double bonds from the origin allows a series of furan-based silyloxypolyenes to add to aldehyde carbonyls at the most distant point of the molecule. Denmark's axially chiral bisphosphoramide/SiCl(4) combination catalyzes these scantly precedented, long-range Mukaiyama-type vinylogous aldol reactions efficiently, providing a palette of extended γ-alkylidene butenolide carbinols with complete ω-site selectivity and good to excellent levels of enantioselectivity.
Sigma-Aldrich
Silicon tetrachloride solution, 1.0 M in methylene chloride
Sigma-Aldrich
Silicon tetrachloride, 99.998% trace metals basis
Sigma-Aldrich
Silicon tetrachloride, 99%
Sigma-Aldrich
Silicon tetrachloride, packaged for use in deposition systems