Merck
  • Dynamic heterogeneities, boson peak, and activation volume in glass-forming liquids.
Dynamic heterogeneities, boson peak, and activation volume in glass-forming liquids.
Physical review. E, Statistical, nonlinear, and soft matter physics (2011-07-30)
L Hong, V N Novikov, A P Sokolov
ABSTRACT
There are various arguments and models connecting the characteristic length associated with the boson peak vibrations ξ to the length scale of dynamical heterogeneity L(het). ξ is usually defined as the ratio of the transverse sound velocity to the boson peak frequency. Here we present pressure, temperature, and molecular weight dependencies of ξ, estimated using light scattering, in a few molecular and polymeric glass formers. These dependencies are compared with respective dependencies of the activation volume ΔV(#) in the same materials. Good agreement is found for the pressure and molecular weight dependencies of ξ and ΔV(#) measured at the glass transition temperature T(g). These results provide more evidence for a possible relationship between the sensitivity of structural relaxation to density (activation volume) and the heterogeneity volume. However, contrary to the expectations for L(het), ξ does not decrease with temperature above T(g) in most of the studied materials. The temperature dependence of ξ is compared to that of L(het) in glycerol and orthoterphenyl (OTP) estimated from literature data. The analysis shows a clear difference in the behavior of ξ(T) and ΔV(#)(T) at temperatures above T(g), although ΔV(#)(T)(1/3) and L(het)(T) have similar temperature dependence. Possible reasons for the observed difference are discussed.
Sigma-Aldrich
Polypropylene glycol, P 2,000
Sigma-Aldrich
Poly(propylene glycol), average Mn ~4,000
Sigma-Aldrich
Poly(propylene glycol), average Mn ~2,700
Sigma-Aldrich
Poly(propylene glycol), average Mn ~1,000
Sigma-Aldrich
Poly(propylene glycol), average Mn ~2,000
Sigma-Aldrich
Polypropylene glycol, P 400
Sigma-Aldrich
Poly(propylene glycol), average Mn ~425
Sigma-Aldrich
Poly(propylene glycol), average Mn ~725