Merck
  • Probing autoinducer-2 based quorum sensing: the biological consequences of molecules unable to traverse equilibrium states.
Probing autoinducer-2 based quorum sensing: the biological consequences of molecules unable to traverse equilibrium states.
The Journal of organic chemistry (2011-06-18)
Kyoji Tsuchikama, Colin A Lowery, Kim D Janda
ABSTRACT
Bacteria have developed a cell-to-cell communication system, termed quorum sensing (QS), which allows for the population-dependent coordination of their behavior via the exchange of chemical signals. Autoinducer-2 (AI-2), a class of QS signals derived from 4,5-dihydroxy-2,3-pentandione (DPD), has been revealed as a universal signaling molecule in a variety of bacterial species. In spite of considerable interest, the study of putative AI-2 based QS systems remains a challenging topic in part due to the rapid interconversion between the linear and cyclic forms of DPD. Herein, we report the design and development of efficient syntheses of carbocyclic analogues of DPD, which are locked in the cyclic form. The synthetic analogues were evaluated for the modulation of AI-2-based QS in Vibrio harveyi and Salmonella typhimurium. No agonists were uncovered in either V. harveyi or S. typhimurium assay, whereas weak to moderate antagonists were found against V. harveyi. On the basis of NMR analyses and DFT calculations, the heterocyclic oxygen atom within DPD appears necessary to promote hydration at the C3 position of cyclic DPD to afford the active tetrahydroxy species. These results also shed light on the interaction between the heterocyclic oxygen atom and receptor proteins as well as the importance of the linear form and dynamic equilibrium of DPD as crucial requirements for activation of AI-2 based QS circuits.
Product Description
Supelco
Acetylacetone, analytical standard
Sigma-Aldrich
Acetylacetone, produced by Wacker Chemie AG, Burghausen, Germany, ≥99.5% (GC)
Sigma-Aldrich
Acetylacetone, JIS special grade, ≥99%
Sigma-Aldrich
Acetylacetone, ReagentPlus®, ≥99%