Merck
  • Comparison of manganese oxide nanoparticles and manganese sulfate with regard to oxidative stress, uptake and apoptosis in alveolar epithelial cells.
Comparison of manganese oxide nanoparticles and manganese sulfate with regard to oxidative stress, uptake and apoptosis in alveolar epithelial cells.
Toxicology letters (2011-06-15)
Ramon Frick, Björn Müller-Edenborn, Andreas Schlicker, Barbara Rothen-Rutishauser, David O Raemy, Detlef Günther, Bodo Hattendorf, Wendelin Stark, Beatrice Beck-Schimmer
ABSTRACT
Due to their physicochemical characteristics, metal oxide nanoparticles (NPs) interact differently with cells compared to larger particles or soluble metals. Oxidative stress and cellular metal uptake were quantified in rat type II alveolar epithelial cells in culture exposed to three different NPs: manganese(II,III) oxide nanoparticles (Mn(3)O(4)-NPs), the soluble manganese sulfate (Mn-salt) at corresponding equivalent doses, titanium dioxide (TiO(2)-NPs) and cerium dioxide nanoparticles (CeO(2)-NPs). In the presence of reactive oxygen species an increased apoptosis rate was hypothesized. Oxidative stress was assessed by detection of fluorescently labeled reactive oxygen species and by measuring intracellular oxidized glutathione. Catalytic activity was determined by measuring catalyst-dependent oxidation of thiols (DTT-assay) in a cell free environment. Inductively coupled plasma mass spectrometry was used to quantify cellular metal uptake. Apoptosis rate was determined assessing the activity of caspase-3 and by fluorescence microscopic quantification of apoptotic nuclei. Reactive oxygen species were mainly generated in cells treated with Mn(3)O(4)-NPs. Only Mn(3)O(4)-NPs oxidized intracellular glutathione. Catalytic activity could be exclusively shown for Mn(3)O(4)-NPs. Cellular metal uptake was similar for all particles, whereas Mn-salt could hardly be detected within the cell. Apoptosis was induced by both, Mn(3)O(4)-NPs and Mn-salt. The combination of catalytic activity and capability of passing the cell membrane contributes to the toxicity of Mn(3)O(4)-NPs. Apoptosis of samples treated with Mn-salt is triggered by different, potentially extracellular mechanisms.
Sigma-Aldrich
Manganese(II) sulfate monohydrate, puriss., meets analytical specification of Ph. Eur, BP, USP, FCC, 99-100.5% (calc. for dried substance)
Pricing and availability is not currently available.
Sigma-Aldrich
Manganese(II) sulfate monohydrate, ReagentPlus®, ≥99%
Pricing and availability is not currently available.
Sigma-Aldrich
Manganese(II) sulfate monohydrate, ACS reagent, ≥98%
Pricing and availability is not currently available.
Sigma-Aldrich
Manganese(II) sulfate monohydrate, BioReagent, suitable for plant cell culture, suitable for cell culture
Pricing and availability is not currently available.
Sigma-Aldrich
Manganese sulfate monohydrate, meets USP testing specifications
Pricing and availability is not currently available.