Merck
  • Oxidative stability of cooked, frozen, reheated beef patties: effect of antioxidants.
Oxidative stability of cooked, frozen, reheated beef patties: effect of antioxidants.
Journal of the science of food and agriculture (2011-01-22)
Paola Colindres, M Susan Brewer
ABSTRACT
The effect of selected antioxidants (grape seed extract (GS), oleoresin rosemary (OR), water-soluble oregano extract (WO), propyl gallate (PG), butylated hydroxyanisole (BHA)), butylated hydroxytoluene (BHT)) on sensory, color and oxidative stability of cooked, frozen, reheated ground beef patties was evaluated. Beef lean and trim were ground; antioxidants and salt were added. Patties were cooked (71 °C), overwrapped in commercial polyvinyl chloride film, and stored frozen (-18 °C), then evaluated monthly for 6 months. Flavor, odor and color were determined using a descriptive panel. Instrumental color was determined by a spectrocolorimeter. Lipid oxidation was determined using thiobarbituric acid reactive substances (TBARS). After 6 months of storage, PG and GS samples had lower rancid odor scores and TBARS than controls. Control samples and those containing BHT did not differ statistically in sensory grassy or rancid odor, indicating that they were the most oxidized. TBARS correlated with grassy, rancid, cardboard and beef odors during the 6-month storage period. Based on TBARS, the order of effectiveness of the antioxidants was PG and GS > OR > BHA > WO and BHT > control. TBARS were well correlated with sensory evaluations of odor and flavor. Antioxidants also protected a* values during storage.
Product Description
Supelco
Propyl gallate, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Propyl gallate, powder
Sigma-Aldrich
Propyl gallate, ≥98%, FCC
Sigma-Aldrich
Propyl gallate, antioxidant, ≥98.0% (HPLC)
Sigma-Aldrich
Propyl gallate, for microscopy, ≥98.0% (HPLC)