Merck
  • Transformation of unipolar single-walled carbon nanotube field effect transistors to ambipolar induced by polystyrene nanosphere assembly.
Transformation of unipolar single-walled carbon nanotube field effect transistors to ambipolar induced by polystyrene nanosphere assembly.
ACS nano (2009-02-12)
Di Wei, Yan Zhang, Yang Yang, David G Hasko, DaPing Chu, Ken B K Teo, Gehan A J Amaratunga, William I Milne
ABSTRACT
We have fabricated single-walled carbon nanotube (SWNT) field effect transistors (FETs) with molybdenum source and drain contacts. Normally, these devices operate only as p-channel transistors, however, after polystyrene latex nanospheres were attached to the nanotubes close to the contacts, they changed to ambipolar operation. This strategy provides a simple method to modify the electrical behavior of unipolar SWNT-FETs by influencing the gate-channel electric field distribution and offset charge, so enabling complementary circuits to be fabricated.
Sigma-Aldrich
Polystyrene, average Mw ~192,000
Sigma-Aldrich
Polystyrene, average Mw ~400,000 by GPC
Sigma-Aldrich
Polystyrene, average Mw ~350,000, average Mn ~170,000
Sigma-Aldrich
Polystyrene, average Mw ~280,000 by GPC