Merck
On the mechanism of floating and sliding of liquid marbles.
Chemphyschem : a European journal of chemical physics and physical chemistry (2009-01-30)
Edward Bormashenko, Yelena Bormashenko, Albina Musin, Zahava Barkay
ABSTRACT
The mechanisms of floating and sliding of liquid marbles are studied. Liquid marbles containing CaCl(2) and marbles containing NaOH water solutions float on water containing Na(2)CO(3) and an alcoholic solution of phenolphthalein with no chemical reaction. Sliding of liquid marbles, consisting of NaOH water solutions, on polymer substrates coated with phenolphthalein is studied as well. No chemical reaction is observed. These observations supply direct experimental evidence for the suggestion that interfaces are separated by an air layer when marbles roll on solid substrates. It is concluded that a liquid marble rests on hydrophobic particles coating the liquid. In contrast, drops containing an NaOH water solution sliding on superhydrophobic surfaces coated with phenolphthalein leave a colored trace. The mechanism of low-friction sliding of drops deposited on superhydrophobic surfaces and liquid marbles turns out to be quite different: there is no direct contact between liquid and solid in the case of marbles' motion.
Sigma-Aldrich
Phenolphthalein solution, 0.04 % (w/v), pH 7.8-10.0
Pricing and availability is not currently available.
Sigma-Aldrich
Phenolphthalein solution, 0.1 % (w/v), pH 7.8-10.0
Pricing and availability is not currently available.
Sigma-Aldrich
Phenolphthalein, JIS special grade, ≥98.0%
Pricing and availability is not currently available.
Sigma-Aldrich
Phenolphthalein solution, 1 % (w/v), pH 7.8-10.0
Pricing and availability is not currently available.
Sigma-Aldrich
Phenolphthalein, SAJ first grade, ≥97.0%
Pricing and availability is not currently available.
Sigma-Aldrich
Phenolphthalein, ACS reagent
Pricing and availability is not currently available.
Sigma-Aldrich
Phenolphthalein solution, 0.5 wt. % in ethanol: water (1:1)
Pricing and availability is not currently available.