Merck
  • Click-type reaction of aromatic polyamines for improvement of thermal and optoelectronic properties.
Click-type reaction of aromatic polyamines for improvement of thermal and optoelectronic properties.
Journal of the American Chemical Society (2008-10-07)
Tsuyoshi Michinobu
ABSTRACT
A quantitative addition reaction between aromatic amino-substituted alkynes and tetracyanoethylene (TCNE), yielding donor-substituted tetracyanobutadiene chromophores, was for the first time employed as a click-type reaction to improve the thermal and optoelectronic properties of aromatic polyamines. The first reduction potentials or the LUMO levels of the aromatic polyamines were found to linearly decrease by the stepwise TCNE addition.
Sigma-Aldrich
Tetracyanoethylene, 96%
Pricing and availability is not currently available.