Merck
  • Electrophile-induced dearomatizing spirocyclization of N-arylisonicotinamides: a route to spirocyclic piperidines.
Electrophile-induced dearomatizing spirocyclization of N-arylisonicotinamides: a route to spirocyclic piperidines.
Organic letters (2008-06-17)
Gareth Arnott, Heloise Brice, Jonathan Clayden, Emma Blaney
ABSTRACT
Treatment of N-arylisonicotinamides with trifluoromethanesulfonic anhydride triggers intramolecular nucleophilic attack of the aryl ring on the 4-position of the pyridinium intermediate. The products are spirocyclic dihydropyridines which can be converted to valuable spirocyclic piperidines related to biologically active molecules such as MK-677.
Sigma-Aldrich
Isonicotinamide, ReagentPlus®, 99%
Pricing and availability is not currently available.