Merck
New polyphenolic beta-lactams with antioxidant activity.
Chemistry & biodiversity (2008-05-22)
Gianfranco Cainelli, Cristina Angeloni, Rinaldo Cervellati, Paola Galletti, Daria Giacomini, Silvana Hrelia, Riccardo Sinisi
ABSTRACT
Novel 4-alkylidene-beta-lactams with a polyphenolic side chain were synthesized and evaluated as radical scavengers. We have undertaken a detailed study of the antioxidant activity in vitro with chemical and biological testing of the new beta-lactams and of the corresponding methyl polyhydroxy benzoates. Antioxidant activity of beta-lactams and methyl benzoates was measured with the Briggs-Rauscher oscillating reaction, the TEAC (Trolox( Equivalent Antioxidant Capacity) assay, and as ability to inhibit ROS (=Reactive Oxygen Species) production on myoblast H9c2 cells. The results were discussed with regard to mechanism and correlated with structural parameters.
Product Description
Supelco
Methyl benzoate, analytical standard
Sigma-Aldrich
Methyl benzoate, natural, ≥98%, FCC, FG
Sigma-Aldrich
Methyl benzoate, 99%
Sigma-Aldrich
Methyl benzoate, ≥98%, FCC, FG