Merck
  • Continuing efforts on the improvement of Beckmann rearrangement of indanone oxime.
Continuing efforts on the improvement of Beckmann rearrangement of indanone oxime.
Bioorganic & medicinal chemistry letters (2006-10-28)
Yasuhiro Torisawa, Takao Nishi, Jun-ichi Minamikawa
ABSTRACT
Continuing search for beneficial additive toward the Beckmann rearrangement (BR) of indanone oxime has revealed that common Lewis acid catalyst in methanesulfonyl chloride (MsCl) showed increasing efficiency in this ionic rearrangement. The new protocol with MsCl is superior to the classical phosphorus-based methods such as PPA and Eaton reagent, especially in the reaction of indanone oximes.
Sigma-Aldrich
Methanesulfonyl chloride, purum, ≥98.0% (AT)
Sigma-Aldrich
Methanesulfonyl chloride, ≥99.7%