Merck
  • Synthesis of enantiopure (alphaS,betaS)- or (alphaR,betaS)-beta-amino alcohols by complete regioselective opening of aminoepoxides by organolithium reagents LiAlH4 or LiAlD4.
Synthesis of enantiopure (alphaS,betaS)- or (alphaR,betaS)-beta-amino alcohols by complete regioselective opening of aminoepoxides by organolithium reagents LiAlH4 or LiAlD4.
The Journal of organic chemistry (2006-08-12)
José M Concellón, Pablo L Bernad, Virginia del Solar, José Ramón Suárez, Santiago García-Granda, M Rosario Díaz
ABSTRACT
The reaction of chiral (2R,1'S)- or (2S,1'S)-2-(1-aminoalkyl)epoxides, 1 or 2 with a variety of organolithium compounds to obtain the corresponding (alphaS,betaS)- or (alphaR,betaS)- beta-amino alcohols in enantiopure form is reported. In both cases, the opening of the oxirane ring at C-3 proceeded with total regioselectivity. Moreover, the ring opening of aminoepoxides 1 or 2 by hydride (utilizing LiAlH4) to obtain the corresponding (2S,3S)- or (2R,3S)-3-aminoalkan-2-ols is also described. The reaction of 1 or 2 with LiAlD4 in place of LiAlH4 gave the corresponding (2S,3S)- or (2R,3S)-3-amino-1-deuterioalkan-2-ols.
Sigma-Aldrich
Lithium aluminum hydride, hydrogen-storage grade
Sigma-Aldrich
Lithium aluminum hydride, pellets, reagent grade, 95%
Sigma-Aldrich
Lithium aluminum hydride, powder, reagent grade, 95%
Sigma-Aldrich
Lithium aluminum hydride, ≥97.0% (gas-volumetric)
Sigma-Aldrich
Lithium aluminum hydride solution, 2.0 M in THF
Sigma-Aldrich
Lithium aluminum hydride solution, 0.5 M in 2-methoxyethyl ether
Sigma-Aldrich
Lithium aluminum hydride solution, 1.0 M in diethyl ether
Sigma-Aldrich
Lithium aluminum hydride solution, 1.0 M in THF
Sigma-Aldrich
Lithium aluminum hydride solution, 2.3 M in 2-methyltetrahydrofuran