Merck
  • Periodontal debridement with povidone-iodine in periodontal treatment: short-term clinical and biochemical observations.
Periodontal debridement with povidone-iodine in periodontal treatment: short-term clinical and biochemical observations.
Journal of periodontology (2006-03-04)
Guilherme M Zanatta, Sandro Bittencourt, Francisco H Nociti, Enilson A Sallum, Antonio W Sallum, Marcio Z Casati
ABSTRACT
The aim of the present study was to evaluate the clinical effects of one-stage periodontal debridement with an ultrasonic instrument, associated with 0.5% povidone (pvp)-iodine irrigation in patients with chronic periodontitis. Forty-five patients were randomly assigned into three groups: the control group (CG) received quadrant root planing at 1-week intervals over four consecutive sessions; the periodontal debridement plus pvp-iodine group (PD-PIG) received a 45-minute full-mouth debridement with an ultrasonic instrument, associated with 0.5% pvp-iodine irrigation; and the periodontal debridement group (PDG) received a 45-minute full-mouth periodontal debridement with an ultrasonic instrument, associated with NaCl irrigation. At the 3-month evaluation, the mean probing depth (PD) reduction in CG was 2.51+/-0.52 mm, 2.53+/-0.50 mm in PD-PIG, and 2.58+/-0.60 mm in PDG (P<0.05). The clinical attachment level (CAL) analysis showed a statistically significant gain in all groups compared to baseline (1.87+/-0.56 mm [CG], 1.94+/-0.70 mm [PD-PIG], and 1.99+/-0.92 mm [PDG]). Intergroup analysis of PD and CAL at 1 and 3 months showed no differences (P>0.05). The N-benzoyl-L-arginine-p-nitroanilide (BAPNA) test showed a significant reduction in trypsin activity only during the first month (P<0.05); at 3 months there were no differences compared to baseline (P=0.80). This study provides no evidence that pvp-iodine is effective as an adjunct for one-stage periodontal debridement.
Sigma-Aldrich
Nα-Benzoyl-DL-arginine 4-nitroanilide hydrochloride, ≥98%
Pricing and availability is not currently available.