Merck
  • Selective tert-butyl ester deprotection in the presence of acid labile protecting groups with use of ZnBr2.
Selective tert-butyl ester deprotection in the presence of acid labile protecting groups with use of ZnBr2.
The Journal of organic chemistry (2004-09-18)
Ramesh Kaul, Yann Brouillette, Zohreh Sajjadi, Karl A Hansford, William D Lubell
ABSTRACT
Chemoselective hydrolysis of tert-butyl esters in the presence of other acid-labile groups has been explored by employing alpha-amino esters and ZnBr(2) in DCM. Although N-Boc and N-trityl groups were found to be labile, PhF protected amines were compatible with these Lewis acid deprotection conditions such that a variety of N-(PhF)amino acids were prepared in good yields from their corresponding tert-butyl esters.
Sigma-Aldrich
Zinc bromide, 99.999% trace metals basis
Sigma-Aldrich
Zinc bromide, AnhydroBeads, −10 mesh, 99.999% trace metals basis