Merck
  • Cytosine-substituted metalloporphyrins: receptors for recognition of nucleotides in ion-selective electrodes.
Cytosine-substituted metalloporphyrins: receptors for recognition of nucleotides in ion-selective electrodes.
Organic & biomolecular chemistry (2004-04-06)
Vladimír Král, Tatiana V Shishkanova, Jonathan L Sessler, Christopher T Brown
ABSTRACT
A new series of cytosine-substituted metalloporphyrin conjugates (containing Co(II) and Zn(II) as the coordinated metals) were designed and investigated as nucleotide receptors in PVC-membrane-based ion-selective electrodes under neutral conditions. The potentiometric results indicate that these systems, in particular the Co(II)-containing complex, may be potentially useful sensors for complementary nucleotide substrates in the presence of 10 mol% tridodecylmethylammonium chloride (K(Pot.)(5'-GMP/5'-AMP)= 0.045).
Sigma-Aldrich
Tridodecylmethylammonium iodide, 97%
Supelco
Tridodecylmethylammonium chloride, Selectophore, for ion-selective electrodes
Sigma-Aldrich
Tridodecylmethylammonium chloride, 98%