Merck
  • A novel tankyrase inhibitor decreases canonical Wnt signaling in colon carcinoma cells and reduces tumor growth in conditional APC mutant mice.
A novel tankyrase inhibitor decreases canonical Wnt signaling in colon carcinoma cells and reduces tumor growth in conditional APC mutant mice.
Cancer research (2012-03-24)
Jo Waaler, Ondrej Machon, Lucie Tumova, Huyen Dinh, Vladimir Korinek, Steven Ray Wilson, Jan Erik Paulsen, Nina Marie Pedersen, Tor J Eide, Olga Machonova, Dietmar Gradl, Andrey Voronkov, Jens Peter von Kries, Stefan Krauss
ABSTRACT
Increased nuclear accumulation of β-catenin, a mediator of canonical Wnt signaling, is found in numerous tumors and is frequently associated with tumor progression and metastasis. Inhibition of Wnt/β-catenin signaling therefore is an attractive strategy for anticancer drugs. In this study, we have identified a novel small molecule inhibitor of the β-catenin signaling pathway, JW55, that functions via inhibition of the PARP domain of tankyrase 1 and tankyrase 2 (TNKS1/2), regulators of the β-catenin destruction complex. Inhibition of TNKS1/2 poly(ADP-ribosyl)ation activity by JW55 led to stabilization of AXIN2, a member of the β-catenin destruction complex, followed by increased degradation of β-catenin. In a dose-dependent manner, JW55 inhibited canonical Wnt signaling in colon carcinoma cells that contained mutations in either the APC (adenomatous polyposis coli) locus or in an allele of β-catenin. In addition, JW55 reduced XWnt8-induced axis duplication in Xenopus embryos and tamoxifen-induced polyposis formation in conditional APC mutant mice. Together, our findings provide a novel chemotype for targeting canonical Wnt/β-catenin signaling through inhibiting the PARP domain of TNKS1/2.
Product Description
Supelco
4-Aminobenzoic acid, analytical standard
Sigma-Aldrich
4-Aminobenzoic acid, purified by sublimation, ≥99%
Sigma-Aldrich
4-Aminobenzoic acid, ReagentPlus®, ≥99%
Sigma-Aldrich
4-Aminobenzoic acid, ReagentPlus®, 99%
Sigma-Aldrich
Tamoxifen, ≥99%