Merck
  • Comparison of high-throughput single-cell RNA-seq methods for ex vivo drug screening.
Comparison of high-throughput single-cell RNA-seq methods for ex vivo drug screening.
NAR genomics and bioinformatics (2024-01-30)
Henrik Gezelius, Anna Pia Enblad, Anders Lundmark, Martin Åberg, Kristin Blom, Jakob Rudfeldt, Amanda Raine, Arja Harila, Verónica Rendo, Merja Heinäniemi, Claes Andersson, Jessica Nordlund
ABSTRACT
Functional precision medicine (FPM) aims to optimize patient-specific drug selection based on the unique characteristics of their cancer cells. Recent advancements in high throughput ex vivo drug profiling have accelerated interest in FPM. Here, we present a proof-of-concept study for an integrated experimental system that incorporates ex vivo treatment response with a single-cell gene expression output enabling barcoding of several drug conditions in one single-cell sequencing experiment. We demonstrate this through a proof-of-concept investigation focusing on the glucocorticoid-resistant acute lymphoblastic leukemia (ALL) E/R+ Reh cell line. Three different single-cell transcriptome sequencing (scRNA-seq) approaches were evaluated, each exhibiting high cell recovery and accurate tagging of distinct drug conditions. Notably, our comprehensive analysis revealed variations in library complexity, sensitivity (gene detection), and differential gene expression detection across the methods. Despite these differences, we identified a substantial transcriptional response to fludarabine, a highly relevant drug for treating high-risk ALL, which was consistently recapitulated by all three methods. These findings highlight the potential of our integrated approach for studying drug responses at the single-cell level and emphasize the importance of method selection in scRNA-seq studies. Finally, our data encompassing 27 327 cells are freely available to extend to future scRNA-seq methodological comparisons.
Sigma-Aldrich
5-Azacytidine, ≥98% (HPLC)
Sigma-Aldrich
MULTI-seq Lipid-Modified Oligos, for Single Cell and Single Nucleus Multiplexing