Merck
  • Ebola virus sequesters IRF3 in viral inclusion bodies to evade host antiviral immunity.
Ebola virus sequesters IRF3 in viral inclusion bodies to evade host antiviral immunity.
eLife (2024-01-29)
Lin Zhu, Jing Jin, Tingting Wang, Yong Hu, Hainan Liu, Ting Gao, Qincai Dong, Yanwen Jin, Ping Li, Zijing Liu, Yi Huang, Xuan Liu, Cheng Cao
ABSTRACT
Viral inclusion bodies (IBs) commonly form during the replication of Ebola virus (EBOV) in infected cells, but their role in viral immune evasion has rarely been explored. Here, we found that interferon regulatory factor 3 (IRF3), but not TANK-binding kinase 1 (TBK1) or IκB kinase epsilon (IKKε), was recruited and sequestered in viral IBs when the cells were infected by EBOV transcription- and replication-competent virus-like particles (trVLPs). Nucleoprotein/virion protein 35 (VP35)-induced IBs formation was critical for IRF3 recruitment and sequestration, probably through interaction with STING. Consequently, the association of TBK1 and IRF3, which plays a vital role in type I interferon (IFN-I) induction, was blocked by EBOV trVLPs infection. Additionally, IRF3 phosphorylation and nuclear translocation induced by Sendai virus or poly(I:C) stimulation were suppressed by EBOV trVLPs. Furthermore, downregulation of STING significantly attenuated VP35-induced IRF3 accumulation in IBs. Coexpression of the viral proteins by which IB-like structures formed was much more potent in antagonizing IFN-I than expression of the IFN-I antagonist VP35 alone. These results suggested a novel immune evasion mechanism by which EBOV evades host innate immunity.
Sigma-Aldrich
Monoclonal ANTI-FLAG® M2-Peroxidase (HRP) antibody produced in mouse, clone M2, purified immunoglobulin, buffered aqueous glycerol solution
Sigma-Aldrich
Anti-β-Actin−Peroxidase antibody, Mouse monoclonal, clone AC-15, purified from hybridoma cell culture
Sigma-Aldrich
Anti-c-Myc-Peroxidase antibody, Mouse monoclonal, clone 9E10, purified from hybridoma cell culture
Millipore
EZview Red ANTI-FLAG® M2 Affinity Gel, clone M2
Millipore
EZview Red Anti-c-Myc Affinity Gel