Merck
  • Harnessing PROTAC technology to combat stress hormone receptor activation.
Harnessing PROTAC technology to combat stress hormone receptor activation.
Nature communications (2023-12-10)
Mahshid Gazorpak, Karina M Hugentobler, Dominique Paul, Pierre-Luc Germain, Miriam Kretschmer, Iryna Ivanova, Selina Frei, Kei Mathis, Remo Rudolf, Sergio Mompart Barrenechea, Vincent Fischer, Xiaohan Xue, Aleksandra L Ptaszek, Julian Holzinger, Mattia Privitera, Andreas Hierlemann, Onno C Meijer, Robert Konrat, Erick M Carreira, Johannes Bohacek, Katharina Gapp
ABSTRACT
Counteracting the overactivation of glucocorticoid receptors (GR) is an important therapeutic goal in stress-related psychiatry and beyond. The only clinically approved GR antagonist lacks selectivity and induces unwanted side effects. To complement existing tools of small-molecule-based inhibitors, we present a highly potent, catalytically-driven GR degrader, KH-103, based on proteolysis-targeting chimera technology. This selective degrader enables immediate and reversible GR depletion that is independent of genetic manipulation and circumvents transcriptional adaptations to inhibition. KH-103 achieves passive inhibition, preventing agonistic induction of gene expression, and significantly averts the GR's genomic effects compared to two currently available inhibitors. Application in primary-neuron cultures revealed the dependency of a glucocorticoid-induced increase in spontaneous calcium activity on GR. Finally, we present a proof of concept for application in vivo. KH-103 opens opportunities for a more lucid interpretation of GR functions with translational potential.
Sigma-Aldrich
Anti-Mineralocorticoid Receptor Antibody, clone 6G1, clone 6G1, from mouse
Sigma-Aldrich
Goat Anti-Mouse IgG Antibody, (H+L) HRP conjugate, 1 mg/mL, Chemicon®
Sigma-Aldrich
Anti-GAPDH Antibody, from rabbit, purified by affinity chromatography
Sigma-Aldrich
Cycloheximide solution, Ready-Made Solution, microbial, 100 mg/mL in DMSO, Suitable for cell culture
Sigma-Aldrich
Goat Anti-Rabbit IgG Antibody, (H+L) HRP conjugate, 1 mg/mL, Chemicon®