Merck
  • Human Papillomavirus Genome Copy Number Is Maintained by S-Phase Amplification, Genome Loss to the Cytosol during Mitosis, and Degradation in G1 Phase.
Human Papillomavirus Genome Copy Number Is Maintained by S-Phase Amplification, Genome Loss to the Cytosol during Mitosis, and Degradation in G1 Phase.
Journal of virology (2023-02-08)
Malgorzata Bienkowska-Haba, Katarzyna Zwolinska, Timothy Keiffer, Rona S Scott, Martin Sapp
ABSTRACT
The current model of human papillomavirus (HPV) replication is comprised of three modes of replication. Following infectious delivery, the viral genome is amplified during the establishment phase to reach up to some hundred copies per cell. The HPV genome copy number remains constant during the maintenance stage. The differentiation of infected cells induces HPV genome amplification. Using highly sensitive in situ hybridization (DNAscope) and freshly HPV16-infected as well as established HPV16-positive cell lines, we observed that the viral genome is amplified in each S phase of undifferentiated keratinocytes cultured as monolayers. The nuclear viral genome copy number is reset to pre-S-phase levels during mitosis. The majority of the viral genome fails to tether to host chromosomes and is lost to the cytosol. Cytosolic viral genomes gradually decrease during cell cycle progression. The loss of cytosolic genomes is blocked in the presence of NH4Cl or other drugs that interfere with lysosomal acidification, suggesting the involvement of autophagy in viral genome degradation. These observations were also made with HPV31 cell lines obtained from patient samples. Cytosolic viral genomes were not detected in UMSCC47 cells carrying integrated HPV16 DNA. Analyses of organotypic raft cultures derived from keratinocytes harboring episomal HPV16 revealed the presence of cytosolic viral genomes as well. We conclude that HPV maintains viral genome copy numbers by balancing viral genome amplification during S phase with the loss of viral genomes to the cytosol during mitosis. It seems plausible that restrictions to viral genome tethering to mitotic chromosomes reset genome copy numbers in each cell cycle. IMPORTANCE HPV genome maintenance is currently thought to be achieved by regulating the expression and activity of the viral replication factors E1 and E2. In addition, the E8^E2 repressor has been shown to be important for restricting genome copy numbers by competing with E1 and E2 for binding to the viral origin of replication and by recruiting repressor complexes. Here, we demonstrate that the HPV genome is amplified in each S phase. The nuclear genome copy number is reset during mitosis by a failure of the majority of the genomes to tether to mitotic chromosomes. Rather, HPV genomes accumulate in the cytoplasm of freshly divided cells. Cytosolic viral DNA is degraded in G1 in a lysosome-dependent manner, contributing to the genome copy reset. Our data imply that the mode of replication during establishment and maintenance is the same and further suggest that restrictions to genome tethering significantly contribute to viral genome maintenance.
Product Description
Roche
cOmplete, Mini Protease Inhibitor Cocktail, Tablets provided in a glass vial
Sigma-Aldrich
Chloroquine diphosphate salt, powder or crystals, 98.5-101.0% (EP)
Sigma-Aldrich
UM-SCC-47 HPV-16 Positive Squamous Carcinoma Cell Line, suitable in vitro model of H&N carcinoma studies